Assay Details
| Gene Symbol: daf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027988.5 | NP_001023159.1 | 3 - 4 | 297 | 88 | |
| NM_001027989.5 | NP_001023160.1 | 3 - 4 | 294 | 88 | ||
More Information
Set Membership: |
|

daf-1
Genomic Map