Assay Details
| Gene Symbol: csn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067477.8 | NP_499878.3 | 5 - 6 | 946 | 76 | |
| Gene Symbol: Y38C1AA.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379996.1 | NP_001368219.1 | 8 - 9 | 2764 | 76 | |
More Information
Set Membership: |
|

csn-3
Genomic Map