Assay Details
| Gene Symbol: pnp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380002.1 | NP_001368342.1 | 3 - 4 | 400 | 104 | |
| NM_001380003.1 | NP_001367771.1 | 3 - 4 | 424 | 104 | ||
More Information
Set Membership: |
|

pnp-1
Genomic Map