Assay Details
| Gene Symbol: pnp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380002.1 | NP_001368342.1 | 4 - 5 | 621 | 81 | |
| NM_001380003.1 | NP_001367771.1 | 4 - 5 | 645 | 81 | ||
More Information
Set Membership: |
|

pnp-1
Genomic Map