Assay Details
| Gene Symbol: dsc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067502.6 | NP_499903.3 | 2 - 3 | 445 | 71 | |
| GenBank mRNA | AY428645.1 | - | 2 - 3 | 479 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dsc-4| Gene Symbol: dsc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067502.6 | NP_499903.3 | 2 - 3 | 445 | 71 | |
| GenBank mRNA | AY428645.1 | - | 2 - 3 | 479 | 71 | |
Set Membership: |
|