Assay Details
| Gene Symbol: dsc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067502.6 | NP_499903.3 | 7 - 8 | 1510 | 79 | |
| GenBank mRNA | AY428645.1 | - | 7 - 8 | 1544 | 79 | |
More Information
Set Membership: |
|

dsc-4
Genomic Map