Assay Details
| Gene Symbol: pxmp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067506.5 | NP_499907.1 | 4 - 5 | 481 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pxmp-4| Gene Symbol: pxmp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067506.5 | NP_499907.1 | 4 - 5 | 481 | 90 | |
Set Membership: |
|