Assay Details
| Gene Symbol: cdh-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307480.3 | NP_001294409.1 | 8 - 9 | 1020 | 93 | |
| NM_001307481.3 | NP_001294410.1 | 8 - 9 | 1020 | 93 | ||
| NM_001307482.3 | NP_001294411.1 | 8 - 9 | 1020 | 93 | ||
| NM_001307483.3 | NP_001294412.1 | 8 - 9 | 1020 | 93 | ||
| NM_001307484.3 | NP_001294413.1 | 8 - 9 | 1020 | 93 | ||
| NM_001307485.3 | NP_001294414.1 | 8 - 9 | 1020 | 93 | ||
| NM_001307486.3 | NP_001294415.1 | 8 - 9 | 1020 | 93 | ||
| NM_001307487.3 | NP_001294416.1 | 8 - 9 | 1020 | 93 | ||
More Information
Set Membership: |
|

cdh-8
Genomic Map