Assay Details
| Gene Symbol: cdh-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307480.3 | NP_001294409.1 | 21 - 22 | 4187 | 108 | |
| NM_001307481.3 | NP_001294410.1 | 20 - 21 | 3992 | 108 | ||
| NM_001307482.3 | NP_001294411.1 | 21 - 22 | 4085 | 108 | ||
| NM_001307483.3 | NP_001294412.1 | 22 - 23 | 4280 | 108 | ||
| NM_001307484.3 | NP_001294413.1 | 21 - 22 | 4187 | 108 | ||
| NM_001307485.3 | NP_001294414.1 | 20 - 21 | 3992 | 108 | ||
| NM_001307486.3 | NP_001294415.1 | 21 - 22 | 4085 | 108 | ||
| NM_001307487.3 | NP_001294416.1 | 22 - 23 | 4280 | 108 | ||
More Information
Set Membership: |
|

cdh-8
Genomic Map