Assay Details
| Gene Symbol: cdh-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307430.3 | NP_001294359.1 | 2 - 3 | 265 | 100 | |
| NM_001307480.3 | NP_001294409.1 | 23 - 24 | 4705 | 100 | ||
| NM_001307481.3 | NP_001294410.1 | 22 - 23 | 4510 | 100 | ||
| NM_001307482.3 | NP_001294411.1 | 23 - 24 | 4603 | 100 | ||
| NM_001307483.3 | NP_001294412.1 | 24 - 25 | 4798 | 100 | ||
| NM_001307484.3 | NP_001294413.1 | 24 - 25 | 4660 | 100 | ||
| NM_001307485.3 | NP_001294414.1 | 23 - 24 | 4465 | 100 | ||
| NM_001307486.3 | NP_001294415.1 | 24 - 25 | 4558 | 100 | ||
| NM_001307487.3 | NP_001294416.1 | 25 - 26 | 4753 | 100 | ||
More Information
Set Membership: |
|

cdh-8
Genomic Map