Assay Details
| Gene Symbol: deb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306814.4 | NP_001293743.1 | 16 - 17 | 3100 | 105 | |
| NM_001380305.1 | NP_001368388.1 | 4 - 5 | 526 | 105 | ||
| NM_068703.7 | NP_501104.2 | 15 - 16 | 2860 | 105 | ||
| NM_068704.6 | NP_501105.2 | 6 - 7 | 947 | 105 | ||
More Information
Set Membership: |
|

deb-1
Genomic Map