Assay Details
| Gene Symbol: deb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306816.3 | NP_001293745.1 | 1 - 2 | 135 | 68 | |
| NM_068704.6 | NP_501105.2 | 1 - 2 | 145 | 68 | ||
More Information
Set Membership: |
|

deb-1
Genomic Map