Assay Details
| Gene Symbol: rps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380319.1 | NP_001368426.1 | 1 - 2 | 276 | 77 | |
| NM_001383118.1 | 3 - 4 | 520 | 77 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rps-8| Gene Symbol: rps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380319.1 | NP_001368426.1 | 1 - 2 | 276 | 77 | |
| NM_001383118.1 | 3 - 4 | 520 | 77 | |||
Set Membership: |
|