Assay Details
| Gene Symbol: unc-82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268365.3 | NP_001255294.1 | 2 - 3 | 404 | 121 | |
| NM_001380325.1 | NP_001367367.1 | 2 - 3 | 154 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380325.1 transcripts.
Set Membership: |
|

unc-82
Genomic Map