Assay Details
| Gene Symbol: unc-82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268365.3 | NP_001255294.1 | 19 - 20 | 3612 | 98 | |
| NM_001268366.3 | NP_001255295.1 | 14 - 15 | 2893 | 98 | ||
| NM_001307278.3 | NP_001294207.1 | 14 - 15 | 2944 | 98 | ||
| NM_001380324.1 | NP_001367366.1 | 16 - 17 | 2998 | 98 | ||
| NM_001380325.1 | NP_001367367.1 | 18 - 19 | 3239 | 98 | ||
| NM_068785.7 | NP_501186.5 | 14 - 15 | 2893 | 98 | ||
More Information
Set Membership: |
|

unc-82
Genomic Map