Assay Details
| Gene Symbol: unc-82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307278.3 | NP_001294207.1 | 16 - 17 | 3155 | 88 | |
| NM_068785.7 | NP_501186.5 | 16 - 17 | 3104 | 88 | ||
More Information
Set Membership: |
|

unc-82
Genomic Map