Assay Details
| Gene Symbol: unc-82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268365.3 | NP_001255294.1 | 24 - 25 | 4154 | 86 | |
| NM_001268366.3 | NP_001255295.1 | 19 - 20 | 3360 | 86 | ||
| NM_001268369.2 | NP_001255298.1 | 20 - 21 | 3348 | 86 | ||
| NM_001307278.3 | NP_001294207.1 | 20 - 21 | 3558 | 86 | ||
| NM_001372869.1 | NP_001359805.1 | 20 - 21 | 3348 | 86 | ||
| NM_001380324.1 | NP_001367366.1 | 21 - 22 | 3540 | 86 | ||
| NM_001380325.1 | NP_001367367.1 | 23 - 24 | 3781 | 86 | ||
| NM_001380326.1 | NP_001367365.1 | 1 - 2 | 142 | 86 | ||
| NM_068785.7 | NP_501186.5 | 20 - 21 | 3507 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380326.1 transcripts.
Set Membership: |
|

unc-82
Genomic Map