Assay Details
| Gene Symbol: unc-82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268365.3 | NP_001255294.1 | 7 - 8 | 968 | 75 | |
| NM_001268366.3 | NP_001255295.1 | 2 - 3 | 249 | 75 | ||
| NM_001268369.2 | NP_001255298.1 | 4 - 5 | 354 | 75 | ||
| NM_001307278.3 | NP_001294207.1 | 2 - 3 | 300 | 75 | ||
| NM_001372869.1 | NP_001359805.1 | 4 - 5 | 354 | 75 | ||
| NM_001380324.1 | NP_001367366.1 | 4 - 5 | 354 | 75 | ||
| NM_001380325.1 | NP_001367367.1 | 7 - 8 | 718 | 75 | ||
| NM_068785.7 | NP_501186.5 | 2 - 3 | 249 | 75 | ||
More Information
Set Membership: |
|

unc-82
Genomic Map