Assay Details
| Gene Symbol: egas-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068806.4 | NP_501207.2 | 6 - 7 | 1095 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

egas-4| Gene Symbol: egas-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068806.4 | NP_501207.2 | 6 - 7 | 1095 | 84 | |
Set Membership: |
|