Assay Details
| Gene Symbol: nol-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068832.5 | NP_501233.1 | 7 - 8 | 1993 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nol-10| Gene Symbol: nol-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068832.5 | NP_501233.1 | 7 - 8 | 1993 | 72 | |
Set Membership: |
|