Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307214.3 | NP_001294143.1 | 8 - 9 | 1087 | 55 | |
| NM_001307215.3 | NP_001294144.1 | 8 - 9 | 1087 | 55 | ||
| NM_001307219.4 | NP_001294148.2 | 21 - 22 | 19927 | 55 | ||
| NM_001307220.4 | NP_001294149.2 | 21 - 22 | 19927 | 55 | ||
| NM_001383125.1 | 1 - 2 | 413 | 55 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383125.1 transcripts.
Set Membership: |
|

cla-1
Genomic Map