Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307214.3 | NP_001294143.1 | 10 - 11 | 1888 | 121 | |
| NM_001307215.3 | NP_001294144.1 | 10 - 11 | 1888 | 121 | ||
| NM_001307219.4 | NP_001294148.2 | 23 - 24 | 20728 | 121 | ||
| NM_001307220.4 | NP_001294149.2 | 23 - 24 | 20728 | 121 | ||
| NM_001383125.1 | 3 - 4 | 1214 | 121 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383125.1 transcripts.
Set Membership: |
|

cla-1
Genomic Map