Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307215.3 | NP_001294144.1 | 19 - 20 | 6910 | 70 | |
| NM_001307219.4 | NP_001294148.2 | 32 - 33 | 25750 | 70 | ||
| NM_001383125.1 | 12 - 13 | 6240 | 70 | |||
More Information
Set Membership: |
|

cla-1
Genomic Map