Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307215.3 | NP_001294144.1 | 23 - 24 | 7631 | 66 | |
| NM_001307219.4 | NP_001294148.2 | 36 - 37 | 26471 | 66 | ||
| NM_001383125.1 | 16 - 17 | 6961 | 66 | |||
More Information
Set Membership: |
|

cla-1
Genomic Map