Assay Details
| Gene Symbol: nlp-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068843.6 | NP_501244.1 | 1 - 2 | 88 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nlp-20| Gene Symbol: nlp-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068843.6 | NP_501244.1 | 1 - 2 | 88 | 93 | |
Set Membership: |
|