Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 10 - 11 | 1460 | 119 | |
| NM_001028000.5 | NP_001023171.1 | 8 - 9 | 1292 | 119 | ||
| NM_001028001.5 | NP_001023172.1 | 10 - 11 | 1417 | 119 | ||
| NM_001028002.4 | NP_001023173.1 | 9 - 10 | 1388 | 119 | ||
| NM_001268376.4 | NP_001255305.1 | 9 - 10 | 1322 | 119 | ||
| NM_001380339.1 | NP_001367466.1 | 9 - 10 | 1322 | 119 | ||
| GenBank mRNA | AF168688.1 | - | 9 - 10 | 1388 | 119 | |
| AJ243179.2 | - | 9 - 10 | 1410 | 119 | ||
| AJ243180.2 | - | 10 - 11 | 1483 | 119 | ||
| AJ243181.2 | - | 9 - 10 | 1403 | 119 | ||
| AJ243182.2 | - | 10 - 11 | 1437 | 119 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map