Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 17 - 18 | 3716 | 67 | |
| NM_001028000.5 | NP_001023171.1 | 15 - 16 | 3548 | 67 | ||
| NM_001028001.5 | NP_001023172.1 | 17 - 18 | 3673 | 67 | ||
| NM_001028002.4 | NP_001023173.1 | 16 - 17 | 3644 | 67 | ||
| NM_001268376.4 | NP_001255305.1 | 16 - 17 | 3578 | 67 | ||
| NM_001380339.1 | NP_001367466.1 | 16 - 17 | 3578 | 67 | ||
| GenBank mRNA | AF168688.1 | - | 16 - 17 | 3644 | 67 | |
| AJ243179.2 | - | 16 - 17 | 3666 | 67 | ||
| AJ243180.2 | - | 17 - 18 | 3739 | 67 | ||
| AJ243181.2 | - | 16 - 17 | 3659 | 67 | ||
| AJ243182.2 | - | 17 - 18 | 3693 | 67 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map