Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 24 - 25 | 5991 | 103 | |
| NM_001028000.5 | NP_001023171.1 | 22 - 23 | 5823 | 103 | ||
| NM_001028002.4 | NP_001023173.1 | 23 - 24 | 5919 | 103 | ||
| NM_001268376.4 | NP_001255305.1 | 24 - 25 | 5916 | 103 | ||
| GenBank mRNA | AF168688.1 | - | 23 - 24 | 5919 | 103 | |
| AJ243179.2 | - | 23 - 24 | 5941 | 103 | ||
| AJ243180.2 | - | 24 - 25 | 6014 | 103 | ||
| AJ243181.2 | - | 23 - 24 | 5934 | 103 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map