Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 27 - 28 | 6829 | 114 | |
| NM_001028000.5 | NP_001023171.1 | 25 - 26 | 6661 | 114 | ||
| NM_001028001.5 | NP_001023172.1 | 27 - 28 | 6792 | 114 | ||
| NM_001028002.4 | NP_001023173.1 | 26 - 27 | 6757 | 114 | ||
| NM_001268376.4 | NP_001255305.1 | 27 - 28 | 6754 | 114 | ||
| GenBank mRNA | AF168688.1 | - | 26 - 27 | 6757 | 114 | |
| AJ243179.2 | - | 26 - 27 | 6779 | 114 | ||
| AJ243180.2 | - | 27 - 28 | 6852 | 114 | ||
| AJ243181.2 | - | 26 - 27 | 6772 | 114 | ||
| AJ243182.2 | - | 27 - 28 | 6812 | 114 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map