Assay Details
| Gene Symbol: flp-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068854.4 | NP_501255.1 | 2 - 3 | 346 | 94 | |
| GenBank mRNA | AF042400.1 | - | 2 - 3 | 294 | 94 | |
More Information
Set Membership: |
|

flp-13
Genomic Map