Assay Details
| Gene Symbol: dsb-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268377.2 | NP_001255306.1 | 2 - 3 | 516 | 134 | |
| NM_001268378.1 | NP_001255307.1 | 2 - 3 | 516 | 134 | ||
More Information
Set Membership: |
|

dsb-3
Genomic Map