Assay Details
| Gene Symbol: C33H5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068893.3 | NP_501294.1 | 6 - 7 | 786 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C33H5.2| Gene Symbol: C33H5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068893.3 | NP_501294.1 | 6 - 7 | 786 | 81 | |
Set Membership: |
|