Assay Details
| Gene Symbol: flp-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068905.5 | NP_501306.1 | 2 - 3 | 216 | 74 | |
| GenBank mRNA | AF042396.1 | - | 1 - 2 | 94 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

flp-10| Gene Symbol: flp-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068905.5 | NP_501306.1 | 2 - 3 | 216 | 74 | |
| GenBank mRNA | AF042396.1 | - | 1 - 2 | 94 | 74 | |
Set Membership: |
|