Assay Details
| Gene Symbol: gcy-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068923.7 | NP_501324.2 | 3 - 4 | 854 | 113 | |
| GenBank mRNA | AB201388.1 | - | 3 - 4 | 783 | 113 | |
More Information
Set Membership: |
|

gcy-8
Genomic Map