Assay Details
| Gene Symbol: czw-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068926.6 | NP_501327.1 | 4 - 5 | 1506 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

czw-1| Gene Symbol: czw-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068926.6 | NP_501327.1 | 4 - 5 | 1506 | 82 | |
Set Membership: |
|