Assay Details
| Gene Symbol: arf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068935.6 | NP_501336.1 | 2 - 3 | 166 | 93 | |
| GenBank mRNA | AY519855.1 | - | 2 - 3 | 166 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

arf-3| Gene Symbol: arf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068935.6 | NP_501336.1 | 2 - 3 | 166 | 93 | |
| GenBank mRNA | AY519855.1 | - | 2 - 3 | 166 | 93 | |
Set Membership: |
|