Assay Details
| Gene Symbol: dnc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068943.5 | NP_501344.1 | 2 - 3 | 250 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dnc-4| Gene Symbol: dnc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068943.5 | NP_501344.1 | 2 - 3 | 250 | 94 | |
Set Membership: |
|