Assay Details
| Gene Symbol: sax-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038305.3 | NP_001033394.1 | 10 - 11 | 2992 | 100 | |
| NM_001038306.5 | NP_001033395.1 | 13 - 14 | 3370 | 100 | ||
| NM_001038307.5 | NP_001033396.1 | 13 - 14 | 3380 | 100 | ||
| NM_001038308.4 | NP_001033397.1 | 10 - 11 | 2755 | 100 | ||
| GenBank mRNA | AB206669.1 | - | 13 - 14 | 3307 | 100 | |
| AB206670.1 | - | 10 - 11 | 2755 | 100 | ||
More Information
Set Membership: |
|

sax-7
Genomic Map