Assay Details
| Gene Symbol: sax-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038305.3 | NP_001033394.1 | 12 - 13 | 3484 | 68 | |
| NM_001038306.5 | NP_001033395.1 | 15 - 16 | 3862 | 68 | ||
| NM_001038307.5 | NP_001033396.1 | 15 - 16 | 3872 | 68 | ||
| NM_001038308.4 | NP_001033397.1 | 12 - 13 | 3247 | 68 | ||
| NM_001307232.3 | NP_001294161.1 | 3 - 4 | 562 | 68 | ||
| NM_001307233.3 | NP_001294162.1 | 3 - 4 | 1001 | 68 | ||
| GenBank mRNA | AB206669.1 | - | 15 - 16 | 3799 | 68 | |
| AB206670.1 | - | 12 - 13 | 3247 | 68 | ||
More Information
Set Membership: |
|

sax-7
Genomic Map