Assay Details
| Gene Symbol: sax-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038305.3 | NP_001033394.1 | 13 - 14 | 3597 | 73 | |
| NM_001038306.5 | NP_001033395.1 | 16 - 17 | 3975 | 73 | ||
| NM_001307233.3 | NP_001294162.1 | 4 - 5 | 1114 | 73 | ||
More Information
Set Membership: |
|

sax-7
Genomic Map