Assay Details
| Gene Symbol: sax-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038306.5 | NP_001033395.1 | 3 - 4 | 370 | 67 | |
| NM_001038307.5 | NP_001033396.1 | 3 - 4 | 380 | 67 | ||
| GenBank mRNA | AB206669.1 | - | 3 - 4 | 307 | 67 | |
More Information
Set Membership: |
|

sax-7
Genomic Map