Assay Details
| Gene Symbol: sax-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038305.3 | NP_001033394.1 | 6 - 7 | 1282 | 135 | |
| NM_001038306.5 | NP_001033395.1 | 9 - 10 | 1660 | 135 | ||
| NM_001038307.5 | NP_001033396.1 | 9 - 10 | 1670 | 135 | ||
| NM_001038308.4 | NP_001033397.1 | 6 - 7 | 1045 | 135 | ||
| GenBank mRNA | AB206669.1 | - | 9 - 10 | 1597 | 135 | |
| AB206670.1 | - | 6 - 7 | 1045 | 135 | ||
More Information
Set Membership: |
|

sax-7
Genomic Map