Assay Details
| Gene Symbol: pmk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068964.6 | NP_501365.1 | 3 - 4 | 1091 | 131 | |
| Gene Symbol: faah-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068965.5 | NP_501366.1 | 3 - 4 | 1097 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_068965.5 transcripts.
Set Membership: |
|

pmk-1
Genomic Map