Assay Details
| Gene Symbol: pct-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026141.6 | NP_001021312.1 | 3 - 4 | 359 | 78 | |
| NM_068971.7 | NP_501372.1 | 3 - 4 | 370 | 78 | ||
| NR_131581.1 | 2 - 3 | 292 | 78 | |||
| GenBank mRNA | AF129110.1 | - | 2 - 3 | 316 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001026141.6 transcripts.
Set Membership: |
|

pct-1
Genomic Map