Assay Details
| Gene Symbol: gpx-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028197.6 | NP_001023368.1 | 1 - 2 | 176 | 162 | |
| NR_131582.1 | 1 - 2 | 146 | 162 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gpx-6| Gene Symbol: gpx-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028197.6 | NP_001023368.1 | 1 - 2 | 176 | 162 | |
| NR_131582.1 | 1 - 2 | 146 | 162 | |||
Set Membership: |
|