Assay Details
| Gene Symbol: oat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069041.6 | NP_501442.2 | 2 - 3 | 338 | 127 | |
| GenBank mRNA | AF152095.1 | - | 2 - 3 | 349 | 127 | |
More Information
Set Membership: |
|

oat-1
Genomic Map