Assay Details
| Gene Symbol: oat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069041.6 | NP_501442.2 | 6 - 7 | 768 | 96 | |
| GenBank mRNA | AF152095.1 | - | 6 - 7 | 779 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

oat-1| Gene Symbol: oat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069041.6 | NP_501442.2 | 6 - 7 | 768 | 96 | |
| GenBank mRNA | AF152095.1 | - | 6 - 7 | 779 | 96 | |
Set Membership: |
|