Assay Details
| Gene Symbol: dgk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069075.6 | NP_501476.2 | 4 - 5 | 444 | 123 | |
| NM_069076.5 | NP_501477.2 | 4 - 5 | 445 | 123 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dgk-4| Gene Symbol: dgk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069075.6 | NP_501476.2 | 4 - 5 | 444 | 123 | |
| NM_069076.5 | NP_501477.2 | 4 - 5 | 445 | 123 | ||
Set Membership: |
|