Assay Details
| Gene Symbol: ints-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307283.3 | NP_001294212.1 | 10 - 11 | 2089 | 108 | |
| NM_001307284.2 | NP_001294213.1 | 10 - 11 | 1761 | 108 | ||
More Information
Set Membership: |
|

ints-6
Genomic Map