Assay Details
| Gene Symbol: ints-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307283.3 | NP_001294212.1 | 4 - 5 | 1129 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ints-6| Gene Symbol: ints-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307283.3 | NP_001294212.1 | 4 - 5 | 1129 | 139 | |
Set Membership: |
|